missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99017-50ul
This item is not returnable.
View return policy
Description
PRC1 Polyclonal antibody specifically detects PRC1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| PRC1 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:1500-1:50000, Immunohistochemistry-Paraffin 1:2500-1:10000 | |
| anaphase spindle elongation 1 homolog, ASE1, MGC1671, MGC3669, protein regulating cytokinesis 1, protein regulator of cytokinesis 1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human PRC1. | |
| 50 μg | |
| Cell Cycle and Replication, Mitotic Regulators | |
| 9055 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction