missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRC1 Antibody (16F2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-74508
This item is not returnable.
View return policy
Description
PRC1 Monoclonal specifically detects PRC1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| PRC1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 - 3 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunoprecipitation 1:10 - 1:500 | |
| anaphase spindle elongation 1 homolog, ASE1, MGC1671, MGC3669, protein regulating cytokinesis 1, protein regulator of cytokinesis 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| PRC1 (16F2) | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 16F2 | |
| Unconjugated | |
| O43663 | |
| PRC1 | |
| Recombinant PRC1 protein. | |
| 100 μg | |
| Cell Cycle and Replication, Mitotic Regulators | |
| 9055 | |
| Human, Mouse, Rat | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction