missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRB4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110212
This item is not returnable.
View return policy
Description
Immunogen sequence: SPPGKPQGPP QQEGNKPQGP PPPGKPQGPP PAGGNPQQPQ APPAGKPQGP PPPPQGGRPP RPAQGQQPPQ.
This gene encodes a member of the heterogeneous family of basic, proline-rich, human salivary glycoproteins. The encoded preproprotein undergoes proteolytic processing to generate one or more mature peptides before secretion from the parotid glands. Multiple alleles of this gene exhibiting variations in the length of the tandem repeats have been identified. The reference genome encodes the Small allele. This gene is located in a cluster of closely related salivary proline-rich proteins on chromosome 12. Alternative splicing results in multiple transcript variants encoding different isoforms that may undergo similar proteolytic processing. [provided by RefSeq, Nov 2015].
Specifications
| PRB4 | |
| Polyclonal | |
| Unconjugated | |
| PRB4 | |
| basic salivary proline-rich protein 4; BstNI type basic salivary proline-rich protein 4; parotid o protein; Peptide P-D; Po; PRB4; proline rich protein BstNI subfamily 4; proline-rich protein BstNI subfamily 4; salivary proline-rich protein II-1; salivary proline-rich protein Po | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 5545 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P10163 | |
| PRB4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 178-247 of human PRB4 (NP_002714.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction