missing translation for 'onlineSavingsMsg'
Learn More

Prazosin Hydrochloride, TRC

Product Code. 30137050
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137050 250 mg 250mg
17963555 100 mg 100mg
17902755 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137050 Supplier Toronto Research Chemicals Supplier No. TRCP702325250MG
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19237-84-4
Molecular Formula C19 H21 N5 O4 . Cl H
Molecular Weight (g/mol) 419.86
Synonym Prazosin Hydrochloride, Terazosin Hydrochloride Dihydrate Imp. K (EP) as Hydrochloride, [4-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-1-piperazinyl]-2-furanyl-methanone hydrochloride (1:1), 1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-(2-furanylcarbonyl)-piperazine monohydrochloride, 1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-(2-furoyl)-piperazine monohydrochloride, 2-[4-(2-Furoyl)piperazin-1-yl]-4-amino-6,7-dimethoxyquinazoline hydrochloride, Adversuten, Adverszuten, Alpress LP, CP 12299-1, Deprazolin, Duramipress, Eurex, Furazosin hydrochloride, Hypovase, Hypovasole, Minipress, NSC 292810, Peripress, Pratsiol, Prazosin hydrochloride, Sinetens
IUPAC Name [4-(4-amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl]-(furan-2-yl)methanone;hydrochloride
SMILES Cl.COc1cc2nc(nc(N)c2cc1OC)N3CCN(CC3)C(=O)c4occc4

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C19H21N5O4.ClH/c1-26-15-10-12-13(11-16(15)27-2)21-19(22-17(12)20)24-7-5-23(6-8-24)18(25)14-4-3-9-28-14;/h3-4,9-11H,5-8H2,1-2H3,(H2,20,21,22);1H
Formula Weight 419.136
Chemical Name or Material Prazosin Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.