missing translation for 'onlineSavingsMsg'
Learn More

Praziquantel, MedChemExpress

Product Code. 16410848
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16410848 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16410848 Supplier Medchem Express Supplier No. HYB0244
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Praziquantel is a racemic mixture, which is composed of (R)-Praziquantel and (S)- Praziquantel. Praziquantel is safe and has been used for the research of schistosomiasis.

Praziquantel is a racemic mixture, which is composed of (R)-Praziquantel and (S)- Praziquantel. Praziquantel is safe and has been used for the research of schistosomiasis.

Specifications

Chemical Name or Material Praziquantel
CAS 55268-74-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H24N2O2
Quantity 5 g
Solubility Information DMSO : 50 mg/mL (160.05 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1CN(C(C2CCCCC2)=O)CC3N1CCC4=C3C=CC=C4
Molecular Weight (g/mol) 312.41
Formula Weight 312.41
Percent Purity 99.71%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.