missing translation for 'onlineSavingsMsg'
Learn More

Pravastatin sodium, MedChemExpress

Product Code. 16434267
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16434267

Brand: Medchem Express HYB0165A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pravastatin sodium (CS-514 sodium) is an HMG-CoA reductase inhibitor against sterol synthesis with IC50 of 5.6 μM.

Pravastatin sodium (CS-514 sodium) is an HMG-CoA reductase inhibitor against sterol synthesis with IC50 of 5.6 μM.

Specifications

Chemical Name or Material Pravastatin sodium
Physical Form Powder
CAS 81131-70-6
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C23H35NaO7
For Use With (Application) COVID-19-immunoregulation
Synonym CS-514 sodium
Solubility Information DMSO : 100 mg/mL (223.96 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (111.98 mM)
SMILES O[C@@H](C[C@@H](O)CC(O[Na])=O)CC[C@@H]([C@H](C=C1)C)[C@@](C1=C[C@@H](O)C2)([H])[C@H]2OC([C@@H](C)CC)=O
Molecular Weight (g/mol) 446.51
Formula Weight 446.51
Percent Purity 98.4%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.