missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAT4A/TNRC5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93610-0.02ml
This item is not returnable.
View return policy
Description
PRAT4A/TNRC5 Polyclonal antibody specifically detects PRAT4A/TNRC5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| PRAT4A/TNRC5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| CAG4A, canopy 3 homolog (zebrafish), CTG4A, ERDA5CTG repeat protein 4a, Expanded repeat-domain protein CAG/CTG 5, PRAT4ACAG repeat containing, Protein associated with TLR4, protein associated with Toll-like receptor 4A, protein canopy homolog 3, TNRC5expanded repeat domain, CAG/CTG 5, trinucleotide repeat containing 5, Trinucleotide repeat-containing gene 5 protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 31-278 of human CNPY3 (NP_006577.2). GPSQAGAEENDWVRLPSKCEVCKYVAVELKSAFEETGKTKEVIGTGYGILDQKASGVKYTKSDLRLIEVTETICKRLLDYSLHKERTGSNRFAKGMSETFETLHNLVHKGVKVVMDIPYELWNETSAEVADLKKQCDVLVEEFEEVIEDWYRNHQEEDLTEFLCANHVLKGKDTSCLAEQWSGKKGDTAALGGKKSKKKSSRAKAAGGRSSSSKQRKELGGLEGDPSPEEDEGIQKASPLTHSPPDEL | |
| 0.02 mL | |
| Cell Biology | |
| 10695 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto