missing translation for 'onlineSavingsMsg'
Learn More

Prasugrel, MedChemExpress

Product Code. 16496706
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496706 100 mg 100mg
16441766 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16496706 Supplier Medchem Express Supplier No. HY15284100mg
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Prasugrel (PCR 4099), a thienopyridine and prodrug, inhibits platelet function. Prasugrel is an orally active and potent P2Y12 receptor antagonist, and inhibits ADP-induced platelet aggregation.

Prasugrel (PCR 4099), a thienopyridine and prodrug, inhibits platelet function. Prasugrel is an orally active and potent P2Y12 receptor antagonist, and inhibits ADP-induced platelet aggregation.

Specifications

Chemical Name or Material Prasugrel
CAS 150322-43-3
Color White
Purity Grade Notes Research
Molecular Formula C20H20FNO3S
Quantity 100 mg
Synonym PCR 4099
Solubility Information DMSO : 100 mg/mL (267.78 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES FC1=CC=CC=C1C(C(C2CC2)=O)N3CCC4=C(C3)C=C(S4)OC(C)=O
Molecular Weight (g/mol) 373.44
Formula Weight 373.44
Percent Purity 99.82%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.