missing translation for 'onlineSavingsMsg'
Learn More

Prasugrel Hydroxy Thiolactone, TRC

Product Code. 30421854
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Product Code. Quantity
30421854 1 mg
30426524 5 mg
30412017 10 mg
3 options
This item is not returnable. View return policy

Product Code. 30421854

Brand: Toronto Research Chemicals TRCP7011601MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 947502-66-1
Formula Weight 347.0991
InChI Formula InChI=1S/C18H18FNO3S/c19-14-4-2-1-3-13(14)16(17(22)11-5-6-11)20-8-7-18(23)12(10-20)9-15(21)24-18/h1-4,9,11,16,23H,5-8,10H2
Molecular Formula C18H18FNO3S
Molecular Weight (g/mol) 347.4
Percent Purity >95%
SMILES FC1=CC=CC=C1C(C(C2CC2)=O)N3CCC4(O)C(C3)=CC(S4)=O
Synonym 5-[2-Cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-5,6,7,7a-tetrahydro-7a-hydroxy thieno[3,2-c]pyridin-2(4H)-one
Chemical Name or Material Prasugrel Hydroxy Thiolactone
Grade HPLC

Specifications

Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.