missing translation for 'onlineSavingsMsg'
Learn More

Prasugrel hydrochloride, MedChemExpress

Product Code. 16406716
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16406716

Brand: Medchem Express HY15284A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Prasugrel hydrochloride (PCR 4099 hydrochloride), a thienopyridine and prodrug, inhibits platelet function. Prasugrel hydrochloride is an orally active and potent P2Y12 receptor antagonist, and inhibits ADP-induced platelet aggregation.

Prasugrel hydrochloride (PCR 4099 hydrochloride), a thienopyridine and prodrug, inhibits platelet function. Prasugrel hydrochloride is an orally active and potent P2Y12 receptor antagonist, and inhibits ADP-induced platelet aggregation.

Specifications

Chemical Name or Material Prasugrel hydrochloride
CAS 389574-19-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H21ClFNO3S
Quantity 100 mg
Synonym PCR 4099 hydrochloride
Solubility Information DMSO : 41.67 mg/mL (101.66 mM; Need ultrasonic)
SMILES FC1=CC=CC=C1C(C(C2CC2)=O)N3CCC4=C(C3)C=C(S4)OC(C)=O.Cl
Molecular Weight (g/mol) 409.9
Formula Weight 409.9
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.