missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Prasugrel, 96%

Product Code. 15688735 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15688735 1 g 1g
15698735 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15688735 Supplier Thermo Scientific Chemicals Supplier No. 461480010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 150322-43-3 | C20H20FNO3S | 373.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150322-43-3
Molecular Formula C20H20FNO3S
Molecular Weight (g/mol) 373.44
InChI Key DTGLZDAWLRGWQN-UHFFFAOYSA-N
Synonym prasugrel, effient, efient, prasugrel effient, prasita, 5-2-cyclopropyl-1-2-fluorophenyl-2-oxoethyl-4,5,6,7-tetrahydrothieno 3,2-c pyridin-2-yl acetate, 2-2-acetyloxy-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl-1-cyclopropyl-2-2-fluorophenyl ethanone, effient tn, ethanone, 2-2-acetyloxy-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl-1-cyclopropyl-2-2-fluorophenyl
PubChem CID 6918456
ChEBI CHEBI:87723
IUPAC Name [5-[2-cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-6,7-dihydro-4H-thieno[3,2-c]pyridin-2-yl] acetate
SMILES CC(=O)OC1=CC2=C(S1)CCN(C2)C(C3=CC=CC=C3F)C(=O)C4CC4

Specifications

Melting Point 110.0°C to 121.0°C
Ignition Residue 0.1% max
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 373.44
Percent Purity 98%
Chemical Name or Material Prasugrel

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.