missing translation for 'onlineSavingsMsg'
Learn More

Prasterone (DHEA; Dehydroepiandrosterone) 1.0 mg/ml in Acetonitrile, LoGiCal

Product Code. 30458862
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458862 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458862 Supplier LoGiCal Supplier No. LGCAMP0651.0011

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dehydroisoandrosterone (DHEA)
CAS 53-43-0
Analyte or Component Names Dehydroisoandrosterone (DHEA)
Shipping Condition Room Temperature
Molecular Formula C19 H28 O2
Certifications/Compliance ISO 17034
Synonym 17-Chetovis, 17-Hormoforin, 5,6-Dehydroisoandrosterone, 5,6-Didehydroisoandrosterone, 5-Dehydroepiandrosterone, Androstenolone, Astenile, DHA, DHEA, Deandros, Dehydro-epi-androsterone, Dehydroepiandrosterone, Dehydroisoandrosterone, Diandron, Diandrone, EM 760, GL 701, IM 28, Immunor, Intrarosa, NSC 9896, Prasterone, Psicosterone, Siscelar plus, trans-Dehydroandrosterone, Androst-5-en-17-one, 3-hydroxy-, (3beta)-, Androst-5-en-17-one, 3beta-hydroxy- (8CI), (3beta)-3-Hydroxyandrost-5-en-17-one, 3beta-Hydroxy-Delta5-androsten-17-one, 3beta-Hydroxyandrost-5-en-17-one, 3beta-Hydroxyandrost-5-ene-17-one, 5-Androsten-3beta-ol-17-one, Androst-5-ene-17-one-3beta-ol, Androst-5-ene-3beta-ol-17-one, Delta5-Androsten-3beta-ol-17-one
InChI Formula InChI=1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h3,13-16,20H,4-11H2,1-2H3/t13-,14-,15-,16-,18-,19-/m0/s1
Solution Type Single Solution
SMILES C[C@]12CC[C@H]3[C@@H](CC=C4C[C@@H](O)CC[C@]34C)[C@@H]1CCC2=O
IUPAC Name (3S,8R,9S,10R,13S,14S)-3-hydroxy-10,13-dimethyl-1,2,3,4,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-one
Solvent or Matrix Acetonitrile
Industry Type Clinical Testing
Molecular Weight (g/mol) 288.424
Quantity 1 mL
Formula Weight 288.209 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.