missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium rod, 12.7mm (0.5in) dia, 99.5% (metals basis excluding Ta), Thermo Scientific Chemicals

Catalog Number 11383838
Click to view available options
Quantity:
10 mm
25 mm
50 mm
100 mm
Este artículo no se puede devolver. Vea la política de devoluciones
7440-10-0
Pr
140.91
MFCD00011174
PUDIUYLPXJFUGB-UHFFFAOYSA-N
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
23942
CHEBI:49828
praseodymium
[Pr]
Este artículo no se puede devolver. Vea la política de devoluciones
7440-10-0
Pr
140.91
MFCD00011174
PUDIUYLPXJFUGB-UHFFFAOYSA-N
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
23942
CHEBI:49828
praseodymium
[Pr]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Quantity 10 mm
Percent Purity 99.5%
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.86g/mm
Form Cast Rod
Assay (metals basis excluding Ta)
Chemical Name or Material Praseodymium rod, 12.7mm (0.5 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'