missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) sulfate octahydrate, 99.9% (REO)

Product Code. p-4782093
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11332568

Brand: Thermo Scientific Alfa Aesar 014607.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13510-41-3 | H16O20Pr2S3 | 714.103 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13510-41-3
Molecular Formula H16O20Pr2S3
Molecular Weight (g/mol) 714.103
MDL Number MFCD00149832
InChI Key NIJXLQIGVGZNKP-UHFFFAOYSA-H
Synonym praseodymium sulfate-water 2/3/8, 3so4.2pr.8h2o, praseodymium sulfate, dipraseodymium 3+ ion octahydrate trisulfate, praseodymium iii sulfate octahydrate, reacton, dipraseodymium 3+ octahydrate trisulfate
PubChem CID 25022097
IUPAC Name praseodymium(3+);trisulfate;octahydrate
SMILES O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Pr+3].[Pr+3]

Specifications

Color Green
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.9% (REO)
Formula Weight 714.12 (570.00 Anhydrous)
Odor Odorless
Appearance Green powder
Density 2.827g/cm3
Chemical Name or Material Praseodymium(III) sulfate octahydrate, Rare Earth Oxides Content

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.