missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) sulfate octahydrate, 99.9% (REO)

Product Code. 11332568
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332568 25 g 25g
11322568 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332568 Supplier Thermo Scientific Alfa Aesar Supplier No. 014607.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13510-41-3 | H16O20Pr2S3 | 714.103 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13510-41-3
Molecular Formula H16O20Pr2S3
Molecular Weight (g/mol) 714.103
MDL Number MFCD00149832
InChI Key NIJXLQIGVGZNKP-UHFFFAOYSA-H
Synonym praseodymium sulfate-water 2/3/8, 3so4.2pr.8h2o, praseodymium sulfate, dipraseodymium 3+ ion octahydrate trisulfate, praseodymium iii sulfate octahydrate, reacton, dipraseodymium 3+ octahydrate trisulfate
PubChem CID 25022097
IUPAC Name praseodymium(3+);trisulfate;octahydrate
SMILES O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Pr+3].[Pr+3]

Specifications

Color Green
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.9% (REO)
Formula Weight 714.12 (570.00 Anhydrous)
Odor Odorless
Appearance Green powder
Density 2.827g/cm3
Chemical Name or Material Praseodymium(III) sulfate octahydrate, Rare Earth Oxides Content

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.