missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Praseodymium(III) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
50 g
Descripción
Praseodymium(III) oxide is used as a pigment in glass, optic and ceramic industries. Praseodymium-doped glass called didymium glass is used in welding goggles, since it blocks infra-red radiation. Further, it is used as a dielectric in combination with silicon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Melting Point | 2300°C |
Physical Form | Sintered Lump |
Quantity | 10 g |
Merck Index | 14,7712 |
Solubility Information | Insoluble in water. Moderately soluble in strong mineral acids. |
Formula Weight | 329.83 |
Percent Purity | 99.9% |
Odor | Odorless |
Density | 7.07 g/mL |
Assay | (metals basis) |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'