missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) oxide, 99.9% (metals basis)

Product Code. 11333817
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333817 50 g 50g
11343817 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333817 Supplier Thermo Scientific Alfa Aesar Supplier No. 035663.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12036-32-7 | O3Pr2 | 329.812 g/mol

Praseodymium(III) oxide is used as a pigment in glass, optic and ceramic industries. Praseodymium-doped glass called didymium glass is used in welding goggles, since it blocks infra-red radiation. Further, it is used as a dielectric in combination with silicon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-32-7
Molecular Formula O3Pr2
Molecular Weight (g/mol) 329.812
MDL Number MFCD00011178
InChI Key MMKQUGHLEMYQSG-UHFFFAOYSA-N
Synonym dipraseodymium trioxide, praseodymium iii oxide, praseodymium oxide pr2o3, praseodynium oxide, dipraseodymium 3+ ion trioxidandiide, praseodymium iii oxide trace metals basis 10g
PubChem CID 165911
IUPAC Name oxygen(2-);praseodymium(3+)
SMILES [O-2].[O-2].[O-2].[Pr+3].[Pr+3]

Specifications

Melting Point 2300°C
Physical Form Sintered Lump
Quantity 50 g
Merck Index 14,7712
Solubility Information Insoluble in water. Moderately soluble in strong mineral acids.
Formula Weight 329.83
Percent Purity 99.9%
Odor Odorless
Density 7.07 g/mL
Assay (metals basis)
Chemical Name or Material Praseodymium(III) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.