missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog Number 11338168
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
10361-79-2
MFCD00011176
acmc-20ajts, praseodymium 3+ ion trichloride, praseodymium chloride, anhydrous, praseodymium iii chloride, anhydrous, ampuled under argon trace rare earth metals basis 5g
This item is not returnable. View return policy
10361-79-2
MFCD00011176
acmc-20ajts, praseodymium 3+ ion trichloride, praseodymium chloride, anhydrous, praseodymium iii chloride, anhydrous, ampuled under argon trace rare earth metals basis 5g

Praseodymium(III) chloride is used in producing metal praseodymium and praseodymium compounds,glass,ceramic and coloring.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Boiling Point 1700°C
Melting Point 786°C
Physical Form Powder
Quantity 5 g
Merck Index 14,7712
Solubility Information Soluble in water. (1039g/L) at 13 C.
Formula Weight 247.27
Percent Purity 99.99%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 4.02 g/mL
Assay (REO)
Chemical Name or Material Praseodymium(III) chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'