missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) chloride, ultra dry, 99.95% (REO)

Product Code. 15459105
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459105 25 g 25g
15439105 1 g 1g
15449105 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15459105 Supplier Thermo Scientific Alfa Aesar Supplier No. H37815.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10361-79-2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-79-2
MDL Number MFCD00011176
Synonym acmc-20ajts, praseodymium 3+ ion trichloride, praseodymium chloride, anhydrous, praseodymium iii chloride, anhydrous, ampuled under argon trace rare earth metals basis 5g

Specifications

Boiling Point 1700°C
Melting Point 786°C
Physical Form Powder
Quantity 25 g
Linear Formula PrCl3
Formula Weight 247.27
Percent Purity 99.95%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 4.02 g/mL
Assay (REO)
Chemical Name or Material Praseodymium(III) chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.