missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) carbonate octahydrate, 99.9% (REO)

Product Code. p-4824903
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11342568

Brand: Thermo Scientific Alfa Aesar 014609.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14948-62-0 | C3H16O17Pr2 | 605.959 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14948-62-0
Molecular Formula C3H16O17Pr2
Molecular Weight (g/mol) 605.959
MDL Number MFCD00211307
InChI Key SEHQEMTYQGRHIC-UHFFFAOYSA-H
Synonym praseodymium carbonate octahydrate, 3co3.2pr.8h2o, praseodymium carbonate, praseodymium carbonate-water 2/3/8, dipraseodymium 3+ ion octahydrate trico3, praseodymium iii carbonate octahydrate, reacton, dipraseodymium 3+ octahydrate tricarbonate
PubChem CID 25022096
IUPAC Name praseodymium(3+);tricarbonate;octahydrate
SMILES C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.O.O.O.O.O.O.O.[Pr+3].[Pr+3]

Specifications

Color Green
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.9% (REO)
Formula Weight 605.97 (461.85 Anhydrous)
Odor Odorless
Appearance Green crystalline
Chemical Name or Material Praseodymium(III) carbonate octahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.