missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) bromide, ultra dry, 99.99% (REO)

Product Code. 11378557
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11378557

Brand: Thermo Scientific Alfa Aesar 035714.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13536-53-3

Praseodymium(III) bromide has some of the common uses such as in aircraft engines, motion picture industry, coloring in glasses and enamels, color cubic zirconia to simulate peridot, welder and glass blower goggles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13536-53-3
MDL Number MFCD00049762

Specifications

Melting Point 691°C
Physical Form Powder
Quantity 10 g
Solubility Information It is soluble in water.
Formula Weight 380.63
Percent Purity 99.99%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Praseodymium(III) bromide, Ultra dry

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.