missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Praseodymium(III) acetate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Praseodymium Acetate is used to make color glasses and enamels; when mixed with certain other materials, Praseodymium produces an intense clean yellow color in glass. The other main application is used to manufacturing pigment and ink color.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Crystalline Aggregates |
Quantity | 100 g |
Molecular Formula | C6H9O6Pr |
MDL Number | MFCD00150121 |
Solubility Information | Soluble in water,moderately soluble in strong mineral acids. |
InChI Key | KPXRPRLCONABEG-UHFFFAOYSA-K |
SMILES | [Pr+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
IUPAC Name | praseodymium(3+) triacetate |
Molecular Weight (g/mol) | 318.04 |
Formula Weight | 318.03 (Anhydrous) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'