missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium foil, 0.62mm (0.024 in.) thick, 99.5% (REO), Thermo Scientific Chemicals

Catalog Number 11376587
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
7440-10-0
Pr
140.91
MFCD00011174
PUDIUYLPXJFUGB-UHFFFAOYSA-N
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
23942
CHEBI:49828
praseodymium
[Pr]
This item is not returnable. View return policy
7440-10-0
Pr
140.91
MFCD00011174
PUDIUYLPXJFUGB-UHFFFAOYSA-N
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
23942
CHEBI:49828
praseodymium
[Pr]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Color Gray
Quantity 25 x 25 mm
Percent Purity 99.5%
Odor Odorless
Packaging Mylar under Argon
Weight ≈2.6g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Praseodymium foil, 0.62mm (0.024 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'