missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium foil, 0.25mm (0.01 in.) thick, 99.5% (REO), Thermo Scientific Chemicals

Product Code. 11355769
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
25mm
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11355769 50 x 50 mm Each
11351398 25 x 25 mm 25mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11355769 Supplier Thermo Scientific Alfa Aesar Supplier No. 010305.FI
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-10-0
Molecular Formula Pr
Molecular Weight (g/mol) 140.91
MDL Number MFCD00011174
InChI Key PUDIUYLPXJFUGB-UHFFFAOYSA-N
Synonym unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
PubChem CID 23942
ChEBI CHEBI:49828
IUPAC Name praseodymium
SMILES [Pr]

Specifications

Color Gray
Quantity 50 x 50 mm
Odor Odorless
Packaging Mylar under Argon
Weight ≈1.27g/25x25mm
Form Foil
Percent Purity 99.5%
Assay (REO)
Chemical Name or Material Praseodymium foil, 0.25mm (0.01 in.) thick

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.