missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRAS40 Recombinant Rabbit Monoclonal Antibody (23GB640), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | PRAS40 |
|---|---|
| Clone | 23GB640 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088573
![]() |
Invitrogen™
MA554104 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089505
|
Invitrogen™
MA554105 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRAS40 Recombinant Monoclonal antibody specifically detects PRAS40 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| PRAS40 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q96B36, Q9D1F4 | |
| 292887, 67605, 84335 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB640 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1110012J22Rik; 40 kDa proline-rich AKT substrate; AI227026; AKT1 substrate 1; AKT1 substrate 1 (proline rich); AKT1 substrate 1 (proline-rich); akt1s1; Lobe; Lobel; OTTHUMP00000196756; OTTHUMP00000196760; PKB/Akt substrate 40; Pras; PRAS40; Proline-rich AKT substrate; proline-rich AKT1 substrate 1 | |
| A synthetic peptide of human PRAS40 (150-200AA). | |
| Primary | |
| -20°C | |
| AKT1S1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title