missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRAS40 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534565
This item is not returnable.
View return policy
Description
A suggested positive control is human brain tissue lysate. PA5-34565 can be used with blocking peptide PEP-1608.
The Akt signaling pathway contributes to the regulation of apoptosis after a variety of cell death signals. AKT1S1, also known as PRAS40, is a proline-rich substrate of the kinase AKT1 and is thought to play a role in neuroprotection mediated by nerve growth factor (NGF) after transient focal cerebral ischemia. AKT1S1 is also a substrate and potential regulator of mammalian target of rapamycin (mTOR), a serine/threonine kinase that regulates cell growth and cell cycle, and a negative regulator of autophagy. Treatment with the insulin-like growth factor-1 (IGF1) can induce the phosphorylation of AKT1S1 via the PI3K/AKT signaling pathway in PC12 cells.
Specifications
| PRAS40 | |
| Polyclonal | |
| Unconjugated | |
| AKT1S1 | |
| 1110012J22Rik; 40 kDa proline-rich AKT substrate; AI227026; AKT1 substrate 1; AKT1 substrate 1 (proline rich); AKT1 substrate 1 (proline-rich); akt1s1; Lobe; Lobel; OTTHUMP00000196756; OTTHUMP00000196760; PKB/Akt substrate 40; Pras; PRAS40; Proline-rich AKT substrate; proline-rich AKT1 substrate 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 292887, 67605, 84335 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q96B36, Q9D1F4 | |
| AKT1S1 | |
| A 19 amino acid peptide near the carboxy terminus of human AKT1S1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction