missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAS40 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41221-0.025mg
This item is not returnable.
View return policy
Description
PRAS40 Polyclonal specifically detects PRAS40 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| PRAS40 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| AKT1S1 | |
| Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human AKT1S1. The immunogen is located within amino acids 190 - 240 of AKT1S1. Amino Acid Squence: RTEARSSDEENGPPSSPDL | |
| Affinity Purified | |
| RUO | |
| Primary | |
| At least three isoforms of AKT1S1 are known to exist. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1 - 2 μg/mL, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| 40 kDa, AKT1 substrate 1 (proline-rich), MGC2865, proline-rich AKT1 substrate 1,40 kDa proline-rich AKT substrate | |
| Rabbit | |
| 28 kDa | |
| 0.025 mg | |
| Apoptosis, Autophagy, Cancer, Growth and Development, Lipid and Metabolism, mTOR Pathway, Neuronal Cell Markers, Neuroscience, Tumor Suppressors | |
| 84335 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction