missing translation for 'onlineSavingsMsg'
Learn More

Pranlukast hemihydrate, MedChemExpress

Código de producto. p-200099918
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Tamaño de la unidad:
100mg
50mg
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16478216

Marca: Medchem Express HYB0290A

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

MedChemExpress Pranlukast hemihydrate is a highly potent, selective and competitive antagonist of peptide leukotrienes. Pranlukast inhibits [3H]LTE4, [3H]LTD4, and [3H]LTC4 bindings to lung membranes with Kis of 0.63±0.11, 0.99±0.19, and 5640±680 nM, respectively.

Pranlukast hemihydrate is a highly potent, selective and competitive antagonist of peptide leukotrienes. Pranlukast inhibits [3H]LTE4, [3H]LTD4, and [3H]LTC4 bindings to lung membranes with Kis of 0.63±0.11, 0.99±0.19, and 5640±680 nM, respectively.

Especificaciones

Chemical Name or Material Pranlukast hemihydrate
CAS 150821-03-7
Color White
Purity Grade Notes Research
Molecular Formula C27H23N5O4.1/2H2O
Quantity 100 mg
Synonym ONO-1078 hemihydrate
Solubility Information DMSO : 25 mg/mL (50.97 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.04 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(C1=CC=C(OCCCCC2=CC=CC=C2)C=C1)NC3=C4C(C(C=C(C5=NN=NN5)O4)=O)=CC=C3.[1/2].O
Molecular Weight (g/mol) 490.51
Formula Weight 490.51
Percent Purity 99.93%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Mostrar más Mostrar menos

Research purposes only

Corrección del contenido de un producto

El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.

Título del producto

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.