missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pranlukast hemihydrate, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Description
Pranlukast hemihydrate is a highly potent, selective and competitive antagonist of peptide leukotrienes. Pranlukast inhibits [3H]LTE4, [3H]LTD4, and [3H]LTC4 bindings to lung membranes with Kis of 0.63±0.11, 0.99±0.19, and 5640±680 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | Pranlukast hemihydrate |
| CAS | 150821-03-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C27H23N5O4.1/2H2O |
| Quantity | 100 mg |
| Synonym | ONO-1078 hemihydrate |
| Solubility Information | DMSO : 25 mg/mL (50.97 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.04 mM; ultrasonic and warming and heat to 80°C) |
| SMILES | O=C(C1=CC=C(OCCCCC2=CC=CC=C2)C=C1)NC3=C4C(C(C=C(C5=NN=NN5)O4)=O)=CC=C3.[1/2].O |
| Molecular Weight (g/mol) | 490.51 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction