missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pramocaine hydrochloride, MedChemExpress
Description
Pramocaine hydrochloride decreases the permeability of neuronal membranes to sodium ions, blocking both initiation and conduction of nerve impulses.
Specifications
Specifications
| Chemical Name or Material | Pramocaine hydrochloride |
| CAS | 637-58-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C17H28ClNO3 |
| Quantity | 1 g |
| Synonym | Pramoxine hydrochloride |
| Solubility Information | DMSO : 100 mg/mL (303.16 mM; Need ultrasonic) ∣H2O : 100 mg/mL (303.16 mM; Need ultrasonic) |
| SMILES | CCCCOC1=CC=C(OCCCN2CCOCC2)C=C1.[H]Cl |
| Molecular Weight (g/mol) | 329.86 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?