missing translation for 'onlineSavingsMsg'
Learn More

Pramiracetam, MedChemExpress

Product Code. 16403377
Encompass_Preferred
Click to view available options
Quantity:
50 mg
200 mg
Unit Size:
200mg
50mg
This item is not returnable. View return policy

Product Code. 16403377

Brand: Medchem Express HY17455

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pramiracetam is a nootropic drug derived from piracetam, and is more potent.

Pramiracetam is a nootropic drug derived from piracetam, and is more potent.

Specifications

Chemical Name or Material Pramiracetam
CAS 68497-62-1
Color White
Purity Grade Notes Research
Molecular Formula C14H27N3O2
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (371.22 mM) ∣H2O : ≥ 100 mg/mL (371.22 mM)
SMILES O=C(NCCN(C(C)C)C(C)C)CN1C(CCC1)=O
Molecular Weight (g/mol) 269.38
Formula Weight 269.38
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.