missing translation for 'onlineSavingsMsg'
Learn More

(±)-Pramipexole, TRC

Product Code. 30006388
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
Unit Size:
10mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006388 500 mg 500mg
30004541 50 mg 50mg
30005747 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006388 Supplier Toronto Research Chemicals Supplier No. TRCP700740500MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104617-86-9
Molecular Formula C10H17N3S
Molecular Weight (g/mol) 211.33
Synonym 2-Amino-6-propylamino-4,5,6,7-tetrahydrobenzothiazole, 4,5,6,7-Tetrahydro-N6-propyl-2,6-Benzothiazolediamine;
IUPAC Name 6-N-propyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine
SMILES NC(S1)=NC2=C1C[C@H](NCCC)CC2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H17N3S/c1-2-5-12-7-3-4-8-9(6-7)14-10(11)13-8/h7,12H,2-6H2,1H3,(H2,11,13)
Formula Weight 211.1143
Percent Purity >95%
Chemical Name or Material (±)-Pramipexole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.