missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Pramipexole dihydrochloride
GREENER_CHOICE

Product Code. 18064782
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18064782 10 mg 10mg
18055922 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18064782 Supplier Tocris Bioscience™ Supplier No. 4174/10
  • ONLINE EXCLUSIVE PRICE
    £201.00
    £190.05 / 10mg
    Save £10.95
    5% Off
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selective D3 agonist

Dopamine receptor agonist exhibiting selectivity for the D3 receptor (Ki values are 0.5, 3.3, 3.9 and 3.9 nM for D3, D2S, D2L and D4 receptors respectively). Exhibits negligible affinity for D1 and D5 receptors. Displays activity in the treatment of Parkinson's disease.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104632-25-9
Molecular Formula C10H19Cl2N3S

Specifications

Quantity 10 mg
Formula Weight 284.25
Purity >99%
Molecular Weight (g/mol) 284.243
Chemical Name or Material Pramipexole dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.