missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pramipexole dihydrochloride, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Pramipexole dihydrochloride is a selective and blood-brain barrier (BBB) penetrant dopamine D2-type receptor agonist, with Kis of 2.2 nM, 3.9 nM, 0.5 nM and 1.3 nM for D2-type receptor, D2, D3 and D4 receptors, respectively. Pramipexole dihydrochloride can be used for the research of Parkinson's disease (PD) and restless legs syndrome (RLS).
Specifications
Specifications
| Chemical Name or Material | Pramipexole dihydrochloride |
| Physical Form | Solid |
| CAS | 104632-25-9 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C10H19Cl2N3S |
| For Use With (Application) | Neuroscience-Neurodegeneration |
| Solubility Information | H2O : ≥ 50 mg/mL (175.90 mM) |
| SMILES | NC(S1)=NC2=C1C[C@@H](NCCC)CC2.[H]Cl.[H]Cl |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction