missing translation for 'onlineSavingsMsg'
Learn More

Pramipexole dihydrochloride, MedChemExpress

Product Code. 16447298
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16447298 10 mg 10mg
16403317 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16447298 Supplier Medchem Express Supplier No. HY17355
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Pramipexole dihydrochloride is a selective and blood-brain barrier (BBB) penetrant dopamine D2-type receptor agonist, with Kis of 2.2 nM, 3.9 nM, 0.5 nM and 1.3 nM for D2-type receptor, D2, D3 and D4 receptors, respectively. Pramipexole dihydrochloride can be used for the research of Parkinson's disease (PD) and restless legs syndrome (RLS).

Pramipexole dihydrochloride is a selective and blood-brain barrier (BBB) penetrant dopamine D2-type receptor agonist, with Kis of 2.2 nM, 3.9 nM, 0.5 nM and 1.3 nM for D2-type receptor, D2, D3 and D4 receptors, respectively. Pramipexole dihydrochloride can be used for the research of Parkinson's disease (PD) and restless legs syndrome (RLS).

Specifications

Chemical Name or Material Pramipexole dihydrochloride
Physical Form Solid
CAS 104632-25-9
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C10H19Cl2N3S
For Use With (Application) Neuroscience-Neurodegeneration
Solubility Information H2O : ≥ 50 mg/mL (175.90 mM)
SMILES NC(S1)=NC2=C1C[C@@H](NCCC)CC2.[H]Cl.[H]Cl
Molecular Weight (g/mol) 284.25
Formula Weight 284.25
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.