missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRAMEF7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562463
This item is not returnable.
View return policy
Description
Immunogen sequence: KIIVFCTVPC PRCGIRASYD LEPSHCL Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 44%.
PRAMEF7 gene ontology annotations related to this gene include negative regulation of apoptotic process; negative regulation of cell differentiation; negative regulation of transcription, DNA-templated; positive regulation of cell proliferation.
Specifications
| PRAMEF7 | |
| Polyclonal | |
| Unconjugated | |
| PRAMEF7 | |
| PRAME family member 7; PRAMEF7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 441871 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5VXH5 | |
| PRAMEF7 | |
| Recombinant protein corresponding to Human PRAMEF7. Recombinant protein control fragment (Product #RP-104466). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction