missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAME Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11438-1-AP-150UL
This item is not returnable.
View return policy
Description
PRAME is a 509 amino acid protein belonging to the PRAME family consisting of 7 LRR (leucine-rich) repeats. It is a tumor antigen expressed in human melanomas and is recognized by cytolytic T lymphocytes. It′s overexpression frequently observed in human cancers confers growth or survival advantages by antagonizing retinoic acid receptor (RAR) signaling. PRAME serves as a candidate marker for monitoring childhood acute leukemia. It is expressed in testis and acute leukemias.Specifications
| PRAME | |
| Polyclonal | |
| Unconjugated | |
| PRAME | |
| MAPE, OIP 4, OIP4, Opa interacting protein 4, PRAME | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23532 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P78395 | |
| PRAME | |
| PRAME Fusion Protein Ag1906 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction