missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAME Antibody (Eureka patent anti-PRAME), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28607CL1
This item is not returnable.
View return policy
Description
PRAME Monoclonal antibody specifically detects PRAME in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| PRAME | |
| Monoclonal | |
| CoraFluor 1 | |
| cancer/testis antigen 130, MAPEOIP4CT130, melanoma antigen preferentially expressed in tumors, OIP-4, Opa-interacting protein 4, Opa-interacting protein OIP4, preferentially expressed antigen in melanoma, Preferentially expressed antigen of melanoma | |
| PRAME | |
| 0.1 mL | |
| Cancer | |
| 23532 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Eureka patent anti-PRAME | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction