missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX PRAK Protein, Unactive

Product Code. 17360144
Encompass_Preferred
Click to view available options
Quantity:
250 μg
50 μg
Unit Size:
250µg
50µg
This item is not returnable. View return policy

Product Code. 17360144

Brand: Eurofins DiscoverX 14335

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Unactive, recombinant full-length human PRAK with an N-terminal His-tag, for use in Kinase Assays.

Protein Serine/Threonine Kinase MAPKAPK5 (mitogen-activated protein kinase activated protein kinase 5), also known as p38-regulated/activated protein kinase (PRAK), functions downstream of p38a and p38b in mediating signaling of the p38 pathway. This kinase can be activated in response to cellular stress and proinflammatory cytokines through its phosphorylation by MAP kinases including MAPK1/ERK, MAPK14/p38-alpha, and MAPK11/p38-beta. MAPKAPK5 is localized predominantly in the cytoplasm and is constantly shuttled between the cytoplasm and the nucleus. MAPKAPK5 may play a role in balancing other MAPK pathways because overactivation of MAPKAPK5 can inhibit Ras mediated cell proliferation and gene activation. MAPKAPK5 inhibtion may facilitate cell differentiation, inflammation and cell proliferation.

EMBL AF032437. Protein is R291E with respect to this accession. This conflict is reported inGenBank AI300977, AL556217 and BI495728.

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 50% glycerol, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Liquid at -20°C.
Molecular Weight (g/mol) 55 kDa
Quantity 50 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Full-length
Purity or Quality Grade ≥63%
Protein PRAK, unactive
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.