missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRAK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582529
This item is not returnable.
View return policy
Description
Immunogen sequence: PLHSVNNPIL RKRKLLGTKP KDSVYIHDHE NGAEDSNVAL EKLRDVIAQC ILPQAGKGEN EDEKLNEVMQ EAWKYNRECK LLRDTLQSFS WNGRGFTDKV DRLKLAEIVK QVIEEQTTSH Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 94%.
MAPKAPK5 (PRAK) is activated in response to cellular stress and pro-inflammatory cytokines. Activity of this kinase is regulated by p38 alpha and p38 beta. The protein encoded by PRAK is a member of the serine/threonine kinase family. In response to cellular stress and proinflammatory cytokines, this kinase is activated through its phosphorylation by MAP kinases including MAPK1/ERK, MAPK14/p38-alpha, and MAPK11/p38-beta. In vitro, this kinase phosphorylates heat shock protein HSP27 at its physiologically relevant sites. Two alternately spliced transcript variants of PRAK encoding distinct isoforms have been reported.
Specifications
| PRAK | |
| Polyclonal | |
| Unconjugated | |
| MAPKAPK5 | |
| MAP kinase-activated protein kinase 5; MAPK-activated protein kinase 5; MAPKAP kinase 5; Mapkapk5; MAPKAP-K5; MAPKAPK-5; mitogen-activated protein kinase-activated protein kinase 5; MK5; MK-5; p38-regulated/activated protein kinase; PRAK | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8550 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IW41 | |
| MAPKAPK5 | |
| Recombinant protein corresponding to Human MAPKAPK5. Recombinant protein control fragment (Product #RP-91435). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction