missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-33093
This item is not returnable.
View return policy
Description
PRAK Polyclonal specifically detects PRAK in Human samples. It is validated for Western Blot.
Specifications
| PRAK | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| MAPKAPK5 | |
| Recombinant protein encompassing a sequence within the center region of human PRAK. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.64mg/mL | |
| Western Blot 1:500-1:3000 | |
| EC 2.7.11, MAP kinase-activated protein kinase 5, MAPK-activated protein kinase 5, MAPKAP kinase 5, MAPKAPK-5, mitogen-activated protein kinase-activated protein kinase 5, p38-regulated/activated protein kinase, PRAKEC 2.7.11.1 | |
| Rabbit | |
| 54 kDa | |
| 0.1 mL | |
| Protein Kinase | |
| 8550 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction