missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | PRAF1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228942
|
Novus Biologicals
NBP3-35265-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30226825
|
Novus Biologicals
NBP3-35265-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRAF1 Polyclonal antibody specifically detects PRAF1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western BlotSpecifications
| PRAF1 | |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| DNA-directed RNA polymerase I subunit E, DNA-directed RNA polymerase I subunit RPA49, FLJ13390, FLJ13970, FLJ43482, PAF53RNA polymerase I-associated factor 1, polymerase (RNA) I associated factor 1, polymerase (RNA) I polypeptide E, 53kDa, PRAF1, RNA polymerase I associated factor 53, RNA polymerase I subunit A49, RNA polymerase I-associated factor 53, RP11-405L18.3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-419 of human PRAF1 (NP_071935.1).,, Sequence:, MAAEVLPSARWQYCGAPDGSQRAVLVQFSNGKLQSPGNMRFTLYENKDSTNPRKRNQRILAAETDRLSYVGNNFGTGALKCNTLCRHFVGILNKTSGQMEVYDAELFNMQPLFSDVSVESELALESQTKTYREKMDSCIEAFGTTKQKRALNTRRMNRVGNESLNRAVAKAAETIIDTKGVTALVSDAIHNDLQDDSLYLPPCYDDAAKPEDVYKFEDLLSPAEYEALQSPSEAFRNVTSEEILKMIEENSHCTFVIEALKSLPSDVESRDRQARCIWFLDTLIKFRAHRVVKRKSALGPGVPHIINTKLLKHFTCLTYNNGRLRNLISDSMKAKITAYVIILALHIHDFQIDLTVLQRDLKLSEKRMMEIAKAMRLKISKRRVSVAAGSEEDHKLGTLSLPLPPAQTSDRLAKRRKIT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 64425 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title