missing translation for 'onlineSavingsMsg'
Learn More

Praeruptorin B, MedChemExpress

Product Code. 16438858
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16438858 10 mg 10mg
16433836 25 mg 25mg
16431298 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16438858 Supplier Medchem Express Supplier No. HYN0082

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Praeruptorin B is an inhibitor of sterol regulatory element-binding proteins (SREBPs).

Praeruptorin B is an inhibitor of sterol regulatory element-binding proteins (SREBPs).

Specifications

Chemical Name or Material Praeruptorin B
CAS 73069-28-0
Color White
Purity Grade Notes Research
Molecular Formula C24H26O7
Quantity 10 mg
Synonym Praeruptorin D
Solubility Information DMSO : 25 mg/mL (58.62 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C1C=CC2=CC=C3C([C@H](OC(/C(C)=C\C)=O)[C@H](OC(/C(C)=C\C)=O)C(C)(C)O3)=C2O1
Molecular Weight (g/mol) 426.46
Formula Weight 426.46
Percent Purity 99.88%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.