missing translation for 'onlineSavingsMsg'
Learn More

Pracinostat, MedChemExpress

Product Code. 16478337
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478337 5 mg 5mg
16415648 10 mg 10mg
16401817 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478337 Supplier Medchem Express Supplier No. HY13322
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pracinostat is a potent histone deacetylase (HDAC) inhibitor, with IC50s of 40-140 nM, used for cancer research.

Pracinostat is a potent histone deacetylase (HDAC) inhibitor, with IC50s of 40-140 nM, used for cancer research.

Specifications

Chemical Name or Material Pracinostat
CAS 929016-96-6
Color White
Purity Grade Notes Research
Molecular Formula C20H30N4O2
Quantity 5 mg
Synonym SB939
Solubility Information DMSO : 250 mg/mL (697.39 mM; Need ultrasonic)
SMILES ONC(/C=C/C1=CC=C2N(C(CCCC)=NC2=C1)CCN(CC)CC)=O
Molecular Weight (g/mol) 358.48
Formula Weight 358.48
Percent Purity 98.83%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.