missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ pRABBIT IgG IRES-EmGFP Positive Control Vector

Product Code. 16515651 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16515651 1 Vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 16515651 Supplier Gibco™ Supplier No. A39243
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

pRABBIT IgG IRES-EmGFP Positive Control Vector is a mammalian expression control that expresses a complete, full-length rabbit IgG and EmGFP.

Can be used as a positive expression control for transient expression systems such as the Expi293 Expression System, the ExpiCHO Expression System, the FreeStyle 293 Expression System, and the FreeStyle CHO Expression System.

Features:

  • A mix of pcDNA3.4 plasmids containing the coding sequences for EmGFP and the heavy and light chains of rabbit IgG
  • Optimized mix of IgG light and heavy chain transfection-grade plasmids in a single, ready-to-use tube
  • Expression of rabbit IgG at 450 to 500 mg/L in Expi293F cells
  • Sufficient material for transfection of up to 150 mL of suspension culture in the Expi293, ExpiCHO, and FreeStyle systems

This control is included in the Expi293 GnTI-, Expi293 Inducible and Expi293 Inducible GnTi- expression system kits.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Positive Control DNA Vector
Content And Storage 1 vial (1 mg/mL) pRABBIT IgG IRES-EmGFP Positive Control Vector, store at 2–8°C or long term storage at -20°C

Shipping Condition Dry Ice
Quantity 1 Vial
Grade Transfection Grade

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.