missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PR48 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-41411AF700
This item is not returnable.
View return policy
Description
PR48 Polyclonal antibody specifically detects PR48 in Human samples. It is validated for Western Blot
Specifications
| PR48 | |
| Polyclonal | |
| Western Blot | |
| FLJ60425, NYREN8, NY-REN-8 antigen, PP2A subunit B isoform PR48, PP2A, subunit B, PR48 isoform, PPP2R3L, PPP2R3LY, PR48, protein phosphatase 2 (formerly 2A), regulatory subunit B'', beta, protein phosphatase 2, regulatory subunit B'', beta, Protein phosphatase 2A 48 kDa regulatory subunit, serine/threonine protein phosphatase 2A, 48kDa regulatory subunit B, serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit beta | |
| The immunogen recognized by this antibody maps to a region between residue 350 and 400 of human protein phosphatase 2 (formerly 2A), regulatory subunit B'', beta using the numbering given in entry NP_037371.2 (GeneID 28227). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 28227 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction