missing translation for 'onlineSavingsMsg'
Learn More

PR48 Antibody (2E12), Novus Biologicals™

Product Code. 18349169 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349169 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349169 Supplier Novus Biologicals Supplier No. H00028227M08

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PR48 Monoclonal antibody specifically detects PR48 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PR48
Applications Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation, Sandwich ELISA
Classification Monoclonal
Clone 2E12
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH11180
Gene Alias FLJ60425, NYREN8, NY-REN-8 antigen, PP2A subunit B isoform PR48, PP2A, subunit B, PR48 isoform, PPP2R3L, PPP2R3LY, PR48, protein phosphatase 2 (formerly 2A), regulatory subunit B'', beta, protein phosphatase 2, regulatory subunit B'', beta, Protein phosphatase 2A 48 kDa regulatory subunit, serine/threonine protein phosphatase 2A, 48kDa regulatory subunit B, serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit beta
Host Species Mouse
Immunogen PPP2R3B (AAH11180, 1 a.a. ~ 225 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MIDRIFSGAVTRGRKVQKEGKISYADFVWFLISEEDKKTPTSIEYWFRCMDLDGDGALSMFELEYFYEEQCRRLDSMAIEALPFQDCLCQMLDLVKPRTEGKITLQDLKRCKLANVFFDTFFNIEKYLDHEQKEQISLLRDGDSGGPELSDWEKYAAEEYDILVAEETAGEPWEDGFEAELSPVEQKLSALRSPLAQRPFFEAPSPLGAVDLYEYACGDEDLEPL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 28227
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.