missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PR38B Polyclonal Antibody
GREENER_CHOICE

Product Code. 16650485 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16650485 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16650485 Supplier Invitrogen™ Supplier No. PA5104034

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PRPF38B.

May be required for pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PR38B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PRPF38B
Gene Accession No. Q5VTL8, Q6AXY7, Q80SY5
Gene Alias 1110021E09Rik; AU018955; NET1; pre-mRNA processing factor 38B; pre-mRNA-splicing factor 38B; PRP38 pre-mRNA processing factor 38 (yeast) domain containing B; PRP38 pre-mRNA processing factor 38 domain containing B; Prpf38b; RP11-293A10.1; Sarcoma antigen NY-SAR-27
Gene Symbols PRPF38B
Host Species Rabbit
Immunogen A synthesized peptide derived from human PRPF38B(Accession Q5VTL8), corresponding to amino acid residues K210-S260.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 499691, 55119, 66921
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.