Learn More
PR3 (Proteinase 3) Ab-1, Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ PR3 (Proteinase 3) Ab-1, Mouse Monoclonal Antibody.
£134.64 - £696.00
Specifications
| Antigen | PR3 (Proteinase 3) Ab-1 |
|---|---|
| Clone | MCPR3-2 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Concentration | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Concentration | Price | Quantity & Availability | ||||
|
12689586
|
Epredia
MS-1333-P0 |
100 μL | 200μg/mL | ||||||
|
12609596
|
Epredia
MS-1333-P1 |
500 μL | 200μg/mL | ||||||
|
12679586
|
Epredia
MS-1333-P |
1 mL | 200μg/mL | ||||||
|
12699586
|
Epredia
MS-1333-P1ABX |
100 μL | 1mg/mL | ||||||
|
12619596
|
Epredia
MS-1333-PABX |
200 μL | 1mg/mL | ||||||
Description
Proteinase 3 (PR3), a constituent of azurophil granules of neutrophils (polymorphonuclear cells, PMNs), is the target antigen for most anti-neutrophil cytoplasmic antibodies (c-ANCA) in Wegener's granulomatosis (WG).Host Species: Mouse
Clone: MCPR3-2
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: HMC-1/PR3 cell granule extract
Molecular Weight: 32-40kDa
Positive Control: HMC-1/PR3-S176 cells
Recommended for:
- Western Blotting (Not verified)
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
| PR3 (Proteinase 3) Ab-1 | |
| Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Proteases And Inhibitors | |
| HMC-1/PR3 cell granule extract | |
| Primary |
| MCPR3-2 | |
| Monoclonal | |
| Concentrated | |
| RUO | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.