missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PR3 Monoclonal Antibody (MCPR3-2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA511945
This item is not returnable.
View return policy
Description
MA5-11945 targets Proteinase 3 in IF, FACS and WB applications and shows reactivity with mouse and human samples. The MA5-11945 immunogen is hMC-1/PR3 cell granule extract.
PR3 Monoclonal antibody specifically detects PR3 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.
Specifications
| PR3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P24158, Q61096 | |
| PRTN3 | |
| HMC-1/PR3 cell granule extract | |
| 500 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| MCPR3-2 | |
| Unconjugated | |
| PRTN3 | |
| ACPA; AGP7; azurophil granule protein 7; CANCA; C-ANCA; C-ANCA antigen; leukocyte proteinase 3; MBN; MBT; mPR3; Myeloblastin; neutrophil proteinase 4; NP4; NP-4; P29; PR3; PR-3; Proteinase 3; PRTN3; serine proteinase, neutrophil; wegener autoantigen; Wegener granulomatosis autoantigen | |
| Mouse | |
| Protein G | |
| RUO | |
| 19152, 5657 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction